Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053009.3 | NP_443735.2 | 1, 2 | 1141 | ||
GenBank mRNA | AK147502.1 | - | 1, 2 | 456 | ||
AK147560.1 | - | 1, 2 | 1145 | |||
AK169477.1 | - | 1, 2 | 42 |