Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053009.3 | NP_443735.2 | 11 | 2324 | ||
GenBank mRNA | AK029319.1 | - | 5 | 646 | ||
AK051904.1 | - | 6 | 697 | |||
AK088933.1 | - | 7 | 885 | |||
AK147502.1 | - | 11 | 1639 | |||
AK147560.1 | - | 11 | 2328 | |||
AK169477.1 | - | 11 | 1225 | |||
BC057323.1 | - | 7 | 863 | |||
BC064766.1 | - | 1 | 295 | |||
BC083000.1 | - | 7 | 863 | |||
U05342.1 | - | 9 | 1009 |
Gene Symbol: Cntf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BI739858.1 | - | 614 |