Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145588.1 | NP_663563.1 | 4 | 529 | ||
GenBank mRNA | AF013119.1 | - | 1 | 132 | ||
AK044826.1 | - | 2 | 2079 | |||
AK075922.1 | - | 4 | 520 | |||
AK084310.1 | - | 4 | 517 | |||
AK088033.1 | - | 1 | 105 | |||
AK197204.1 | - | 1 | 19 | |||
BC003427.1 | - | 4 | 529 | 2615715 |