Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025900.2 | NP_080176.2 | 7 | 796 | ||
GenBank mRNA | AK007546.1 | - | 4 | 345 | ||
AK012615.1 | - | 7 | 737 | |||
AK012877.1 | - | 7 | 731 | |||
AK033451.1 | - | 7 | 686 | |||
AK141790.1 | - | 1 | 148 | |||
BC048844.1 | - | 7 | 746 | |||
BC055451.1 | - | 7 | 742 | 4164972 |