Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pygb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153781.1 | NP_722476.1 | 11 | 1363 | ||
GenBank mRNA | AK032769.1 | - | 11 | 1376 | ||
AK035673.1 | - | 11 | 1093 | |||
AK133794.1 | - | 2 | 93 | |||
AK134667.1 | - | 11 | 1373 | |||
AK142716.1 | - | 11 | 1373 | |||
AK147763.1 | - | 11 | 1395 | |||
AK168381.1 | - | 11 | 1374 | |||
AK169052.1 | - | 11 | 1398 | |||
AK169631.1 | - | 11 | 1377 | |||
AK213064.1 | - | 1 | 26 | |||
BC031457.1 | - | 11 | 1371 | |||
BC032209.1 | - | 5 | 518 | |||
BC035283.1 | - | 11 | 1363 | 4947455 |