Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pygb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153781.1 | NP_722476.1 | 19, 20 | 2448 | ||
GenBank mRNA | AK032769.1 | - | 19, 20 | 2461 | ||
AK035673.1 | - | 19, 20 | 2178 | |||
AK133794.1 | - | 10, 11 | 1178 | |||
AK134667.1 | - | 19, 20 | 2458 | |||
AK142716.1 | - | 19, 20 | 2458 | |||
AK147763.1 | - | 19, 20 | 2480 | |||
AK168381.1 | - | 19, 20 | 2460 | |||
AK169052.1 | - | 19, 20 | 2483 | |||
AK184046.1 | - | 2, 3 | 94 | |||
BC031457.1 | - | 19, 20 | 2455 | |||
BC032209.1 | - | 13, 14 | 1603 | |||
BC035283.1 | - | 19, 20 | 2448 | 4947455 |