Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033668.1 | NP_001028840.1 | 20 | 3277 | ||
GenBank mRNA | Z83815.2 | - | 7 | 785 |