Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pygl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133198.2 | NP_573461.2 | 11 | 1463 | ||
GenBank mRNA | AF288783.1 | - | 11 | 1405 | ||
AK140321.1 | - | 11 | 1410 | |||
AK145989.1 | - | 11 | 1464 | |||
AK149491.1 | - | 1638 | ||||
AK167337.1 | - | 11 | 1410 | |||
BC013636.1 | - | 11 | 1397 | 4196237 |