Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Raf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029780.3 | NP_084056.1 | 2 | 578 | ||
GenBank mRNA | AB057655.1 | - | 1 | 123 | ||
AB057663.2 | - | 2 | 348 | |||
AB077145.1 | - | 2 | 348 | |||
AK036317.1 | - | 2 | 465 | |||
AK141745.1 | - | 2 | 579 | |||
BC015273.1 | - | 2 | 457 | 4222659 | ||
BC056439.1 | - | 2 | 497 | |||
BC092040.1 | - | 2 | 443 | 2645313 |