Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Manba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027288.3 | NP_081564.3 | 7 | 977 | ||
GenBank mRNA | AF306557.1 | - | 7 | 981 | ||
AK036505.1 | - | 8 | 1077 | |||
AK047057.1 | - | 7 | 970 | |||
AK142165.1 | - | 7 | 942 | |||
AK154172.1 | - | 7 | 959 | |||
AK166022.1 | - | 7 | 960 | |||
AK166340.1 | - | 7 | 975 | |||
AK170290.1 | - | 7 | 974 | |||
BC031409.1 | - | 7 | 920 | 4952710 |