Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P4HA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000917.3 | NP_000908.2 | 5 | 668 | ||
NM_001017962.2 | NP_001017962.1 | 5 | 668 | |||
NM_001142595.1 | NP_001136067.1 | 6 | 761 | |||
NM_001142596.1 | NP_001136068.1 | 5 | 668 | |||
GenBank mRNA | AK222960.1 | - | 5 | 556 | ||
AK312593.1 | - | 5 | 554 | |||
BC034998.1 | - | 5 | 561 | 4797051 | ||
CD013929.1 | - | 5 | 547 | |||
CD013930.1 | - | 6 | 640 | |||
M24486.1 | - | 5 | 545 | |||
M24487.1 | - | 5 | 545 |