Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253712.1 | NP_001240641.1 | 3 | 435 | ||
NM_001253714.1 | NP_001240643.1 | 2 | 352 | |||
NM_001253716.1 | NP_001240645.1 | 3 | 163 | |||
NM_026322.4 | NP_080598.2 | 8 | 834 | |||
GenBank mRNA | AK002356.1 | - | 3 | 310 | ||
AK009822.1 | - | 3 | 312 | |||
AK018338.1 | - | 3 | 157 | |||
AK030278.1 | - | 3 | 313 | |||
AK049714.1 | - | 3 | 436 | |||
BB652761.1 | - | 3 | 433 | |||
BC089311.1 | - | 8 | 834 | 6412625 | ||
BY282106.1 | - | 3 | 170 | |||
CB953198.1 | - | 2 | 223 |