Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253712.1 | NP_001240641.1 | 2 | 367 | ||
NM_001253715.1 | NP_001240644.1 | 2 | 353 | |||
NM_001253716.1 | NP_001240645.1 | 2 | 95 | |||
NM_026322.4 | NP_080598.2 | 7 | 766 | |||
GenBank mRNA | AK002356.1 | - | 2 | 242 | ||
AK009822.1 | - | 2 | 244 | |||
AK018338.1 | - | 2 | 89 | |||
AK030278.1 | - | 2 | 245 | |||
AK049714.1 | - | 2 | 368 | |||
BB652761.1 | - | 2 | 365 | |||
BC014738.1 | - | 2 | 312 | 4239349 | ||
BC089311.1 | - | 7 | 766 | 6412625 |