Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081412.2 | NP_001074881.1 | 20 | 3557 | ||
GenBank mRNA | AK129482.1 | - | 17 | 1747 | ||
BC002193.1 | - | 3 | 352 | |||
BC048842.1 | - | 8 | 801 | |||
BC060270.1 | - | 20 | 2795 | |||
BC131682.1 | - | 21 | 2962 | 40111128 |