Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080438.2 | NP_536686.2 | 7, 8 | 1009 | ||
GenBank mRNA | AF214575.1 | - | 3, 4 | 411 | ||
AF362764.1 | - | 7, 8 | 927 | |||
AY230204.1 | - | 7, 8 | 948 |