Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027011.2 | NP_081287.1 | 8, 9 | 1540 | ||
GenBank mRNA | AK014363.1 | - | 3, 4 | 324 | ||
BC108361.1 | - | 8, 9 | 1537 | 6312495 |