Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029971.2 | NP_084247.1 | 1 | 298 | ||
GenBank mRNA | AK020723.1 | - | 1 | 298 | ||
AK138363.1 | - | 1 | 301 | |||
BC048534.1 | - | 1 | 298 | 6533154 |