Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144784.3 | NP_659033.1 | 2 | 150 | ||
GenBank mRNA | AJ510150.1 | - | 2 | 142 | ||
AK032097.1 | - | 2 | 121 | |||
AK075593.1 | - | 2 | 120 | |||
AK081715.1 | - | 2 | 150 | |||
AK146241.1 | - | 2 | 105 | |||
AK163402.1 | - | 2 | 154 | |||
AK169771.1 | - | 2 | 143 | |||
BC024429.1 | - | 2 | 153 | |||
BC024763.1 | - | 2 | 122 | 5365469 |
Gene Symbol: Fn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC085240.1 | - | 108 |