Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adarb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024837.2 | NP_001020008.1 | 4 | 655 | ||
NM_130895.3 | NP_570965.2 | 4 | 655 | |||
GenBank mRNA | AF291049.1 | - | 3 | 490 | ||
AF403106.2 | - | 4 | 607 | |||
AF403107.2 | - | 4 | 607 | |||
AF403108.2 | - | 4 | 654 | |||
AF403109.2 | - | 4 | 654 | |||
AF525421.1 | - | 5 | 680 | |||
AK138318.1 | - | 4 | 648 | |||
AK141777.1 | - | 4 | 627 | |||
AK147298.1 | - | 5 | 734 | |||
AY162454.1 | - | 5 | 680 | |||
BC051107.1 | - | 4 | 640 |