Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012026.2 | NP_036156.2 | 6 | 802 | ||
GenBank mRNA | AK173325.1 | - | 6 | 825 | ||
BC022978.1 | - | 6 | 730 | |||
BC057883.1 | - | 6 | 769 | |||
BC075734.1 | - | 6 | 790 | |||
BC098331.1 | - | 6 | 791 | |||
BC138030.1 | - | 6 | 726 | 8861049 | ||
U73199.1 | - | 6 | 802 |