Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078167.2 | NP_001071635.1 | 1 | 570 | ||
NM_173374.4 | NP_775550.2 | 1 | 570 | |||
GenBank mRNA | AK018176.1 | - | 1 | 239 | ||
AK053674.1 | - | 1 | 260 | |||
AK078715.1 | - | 1 | 227 | |||
AK150535.1 | - | 1 | 273 | |||
AK169203.1 | - | 1 | 226 | |||
AK169623.1 | - | 1 | 228 | |||
BC046773.1 | - | 1 | 202 | 5708288 | ||
BC058627.1 | - | 1 | 236 | 6401674 | ||
BY743476.1 | - | 1 | 273 | |||
CF739274.1 | - | 1 | 570 | |||
X66091.1 | - | 1 | 183 |