Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrna5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176844.4 | NP_789814.2 | 5 | 954 | ||
GenBank mRNA | AF204689.1 | - | 4 | 719 | ||
AK080900.1 | - | 2 | 663 | |||
AY574264.1 | - | 5 | 737 | |||
BC106839.2 | - | 5 | 737 | 40045716 | ||
BC106853.2 | - | 5 | 737 | 40046382 | ||
DQ318788.1 | - | 5 | 907 |