Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cds2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291039.1 | NP_001277968.1 | 4 | 284 | ||
NM_001291040.1 | NP_001277969.1 | 4 | 607 | |||
NM_138651.7 | NP_619592.1 | 4 | 607 | |||
GenBank mRNA | AK036328.1 | - | 4 | 458 | ||
AK049257.1 | - | 4 | 285 | |||
AK050589.1 | - | 4 | 515 | |||
AK147541.1 | - | 4 | 499 | |||
AK166001.1 | - | 4 | 458 | |||
AK170888.1 | - | 4 | 496 | |||
AY159802.1 | - | 4 | 530 | |||
BC003852.1 | - | 4 | 513 | 3600825 | ||
BC059776.1 | - | 4 | 410 | |||
BC069879.1 | - | 4 | 486 | 30439069 | ||
CD540005.1 | - | 4 | 282 |