Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052835.3 | NP_443067.1 | 4 | 209 | ||
GenBank mRNA | AK011022.1 | - | 4 | 169 | ||
AK012561.1 | - | 3 | 166 | |||
AK014054.1 | - | 4 | 171 | |||
AK088777.1 | - | 4 | 170 | |||
AK144831.1 | - | 4 | 169 | |||
AK146217.1 | - | 4 | 170 | |||
AK151416.1 | - | 4 | 169 | |||
AK151655.1 | - | 4 | 169 | |||
AK151736.1 | - | 4 | 168 | |||
AK152123.1 | - | 4 | 169 | |||
AK152854.1 | - | 4 | 169 | |||
AK153345.1 | - | 4 | 169 | |||
AK160618.1 | - | 4 | 169 | |||
AK166610.1 | - | 4 | 168 | |||
AK167239.1 | - | 3 | 205 | |||
AK167966.1 | - | 4 | 169 | |||
AK168250.1 | - | 4 | 168 | |||
AK168854.1 | - | 4 | 169 | |||
AK187585.1 | - | 2 | 100 | |||
BC006068.1 | - | 3 | 128 | |||
BC024901.1 | - | 3 | 130 | 4224016 | ||
BC048872.1 | - | 3 | 133 | 5052784 | ||
BC082293.1 | - | 3 | 130 | 6486491 | ||
BC083327.1 | - | 3 | 130 | 5684015 | ||
BC086917.1 | - | 4 | 168 | 6768088 | ||
BC092383.1 | - | 3 | 154 | 5710462 | ||
BC098204.1 | - | 3 | 154 | 6410755 | ||
BC099437.1 | - | 3 | 133 | 30843551 | ||
BY107888.1 | - | 4 | 210 | |||
M93980.1 | - | 4 | 168 | |||
U89405.1 | - | 1 | 38 | |||
X75312.1 | - | 3 | 161 |