Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Highest antigen sequence indentity to the following orthologs: Mouse (91%), Rat (91%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61793. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
AAR2 encodes the homolog of the yeast A1-alpha2 repressin protein that is involved in mRNA splicing. Alternately spliced transcript variants have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AAR2 splicing factor homolog; Protein AAR2 homolog
Gene Aliases: AAR2; C20orf4; CGI-23; PRO0225
UniProt ID: (Human) Q9Y312
Entrez Gene ID: (Human) 25980
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support