Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (69%), Rat (69%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65139. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The structure specific ERCC 1 / XPF endonuclease complex is implicated in the repair of two distinct types of lesions in DNA: NER for UV induced lesions and bulky chemical adducts; and recombination repair of the very genotoxic interstrand cross links. NER mechanism involves dual incisions on both sides of the damage catalyzed by two nucleases. In mammalian cells, XPG cleaves 3' of the DNA lesion while the ERCC1 / XPF complex makes the 5' incision. ERCC1 / XPF probably plays a role in processing partially processed recombination intermediates.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ABC transporter 9 protein; ABC-type oligopeptide transporter ABCB9; ATP-binding cassette sub-family B member 9; ATP-binding cassette transporter 9; ATP-binding cassette, sub-family B (MDR/TAP), member 9; TAP-like protein; TAPL
Gene Aliases: ABCB9; EST122234; KIAA1520; TAPL
UniProt ID: (Human) Q9NP78
Entrez Gene ID: (Human) 23457
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support