Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (81%), Rat (81%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65207. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
ABHD14A play a possible role in granule neuron development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Abhydrolase domain-containing protein 14A; Alpha/beta hydrolase domain-containing protein 14A; Protein ABHD14A
Gene Aliases: ABHD14A; DORZ1; UNQ1913/PRO4373
UniProt ID: (Human) Q9BUJ0
Entrez Gene ID: (Human) 25864
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support