Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Highest antigen sequence indentity to the following orthologs: Mouse (43%), Rat (43%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60768. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acyl-CoA synthetase medium-chain family member 6; Acyl-coenzyme A synthetase ACSM6, mitochondrial; AMP-binding enzyme
Gene Aliases: ACSM6; bA310E22.3; C10orf129
UniProt ID: (Human) Q6P461
Entrez Gene ID: (Human) 142827
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support