Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
It is recommended that the protein be aliquoted for optimal storage.
It is recommended that sterile water (550 µL) be added to the vial to prepare a stock solution of 0.91 mg/mL.
Concentration is measured by UV.
LILRB2 (ILT4) gene is a member of the leukocyte immunoglobulin-like receptor (LIR) family, which is found in a gene cluster at chromosomal region 19q13.4. The encoded protein belongs to the subfamily B class of LIR receptors which contain two or four extracellular immunoglobulin domains, a transmembrane domain, and two to four cytoplasmic immunoreceptor tyrosine-based inhibitory motifs (ITIMs). The receptor is expressed on immune cells where it binds to MHC class I molecules on antigen-presenting cells and transduces a negative signal that inhibits stimulation of an immune response. It is thought to control inflammatory responses and cytotoxicity to help focus the immune response and limit autoreactivity. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD85 antigen-like family member D; CD85d; CD85d antigen; Ig-like transcript 4; ILT-4; Immunoglobulin-like transcript 4; leucocyte Ig-like receptor B2; Leukocyte immunoglobulin-like receptor subfamily B member 2; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 2; LIR-2; LIR2CD85D; MIR-10; MIR10LILRA6; Monocyte/macrophage immunoglobulin-like receptor 10; myeloid inhibitory receptor 10
Gene Aliases: CD85D; ILT-4; ILT4; LILRB2; LIR-2; LIR2; MIR-10; MIR10
UniProt ID: (Human) Q8N423
Entrez Gene ID: (Human) 10288
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support