Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83335. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Clathrin light chain B; clathrin, light chain (Lcb); clathrin, light chain B; clathrin, light polypeptide (Lcb); Lcb
Gene Aliases: CLTB; LCB
UniProt ID: (Human) P09497
Entrez Gene ID: (Human) 1212
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support