Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This peptide corresponds to 14 amino acids near the amino terminus of human Cyclin O.
PEP-0377 can be used as a blocking peptide with polyclonal antibody PA5-20262.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cyclin domain containing; cyclin U; Cyclin-O
Gene Aliases: CCNO; CCNU; CILD29; UDG2
UniProt ID: (Human) P22674
Entrez Gene ID: (Human) 10309
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support