Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This peptide corresponds to 14 amino acids near the carboxy terminus of human FAIM.
PEP-0432 can be used as a blocking peptide with polyclonal antibody PA5-20312.
Plays a role as an inducible effector molecule that mediates Fas resistance produced by surface Ig engagement in B cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FAIM 1; Fas apoptotic inhibitory molecule 1
Gene Aliases: FAIM; FAIM1
UniProt ID: (Human) Q9NVQ4
Entrez Gene ID: (Human) 55179
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support