Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Highest antigen sequence indentity to the following orthologs: Mouse (74%), Rat (74%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51611. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 181, member A; Protein FAM181A
Gene Aliases: C14orf152; FAM181A; HSD-31; HSD31
UniProt ID: (Human) Q8N9Y4
Entrez Gene ID: (Human) 90050
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support