Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (76%), Rat (76%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57453. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 196, member A; Inhibitory synaptic factor 2A; InSyn2
Gene Aliases: C10orf141; FAM196A; INSYN2; INSYN2A
UniProt ID: (Human) Q6ZSG2
Entrez Gene ID: (Human) 642938
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support