Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (65%), Rat (65%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59385. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Transcription cofactor. Negatively regulates transcription activation by catenin beta-1 CTNNB1, perhaps acting by competing with TCF4 for CTNNB1 binding. May play a role in DNA-damage response signaling. Binds specifically to DNA sequences at peri-centromeric chromatin loci.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLYWCH-type zinc finger-containing protein 1
Gene Aliases: FLYWCH1; KIAA1552
UniProt ID: (Human) Q4VC44
Entrez Gene ID: (Human) 84256
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support