Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (73%), Rat (73%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111114. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes coagulation factor XI of the blood coagulation cascade. This protein is present in plasma as a zymogen, which is a unique plasma coagulation enzyme because it exists as a homodimer consisting of two identical polypeptide chains linked by disulfide bonds. During activation of the plasma factor XI, an internal peptide bond is cleaved by factor XIIa (or XII) in each of the two chains, resulting in activated factor XIa, a serine protease composed of two heavy and two light chains held together by disulfide bonds. This activated plasma factor XI triggers the middle phase of the intrisic pathway of blood coagulation by activating factor IX. Defects in this factor lead to Rosenthal syndrome, a blood coagulation abnormality.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coagulation factor XI; FXI; Plasma thromboplastin antecedent; PTA
Gene Aliases: F11; FXI
UniProt ID: (Human) P03951
Entrez Gene ID: (Human) 2160
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support