Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (87%), Rat (87%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57609. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
May function as a transcriptional regulator and a vertebrate homeobox gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Homeobox and leucine zipper protein Homez; Homeodomain leucine zipper-containing factor
Gene Aliases: HOMEZ; KIAA1443
UniProt ID: (Human) Q8IX15
Entrez Gene ID: (Human) 57594
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support