Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54827. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
LLGL1 is a protein that is similar to a tumor suppressor in Drosophila. The protein is part of a cytoskeletal network and is associated with nonmuscle myosin II heavy chain and a kinase that specifically phosphorylates this protein at serine residues. The gene for LLGL1 is located within the Smith-Magenis syndrome region on chromosome 17.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DLG4; Hugl protein; Hugl-1; Human homolog to the D-lgl gene protein; lethal giant larvae homolog 1; lethal giant larvae homolog 1, scribble cell polarity complex component; Lethal(2) giant larvae protein homolog 1; LLGL
Gene Aliases: DLG4; HUGL; HUGL-1; HUGL1; Lgl1; LLGL; LLGL1; Mgl1
UniProt ID: (Human) Q15334
Entrez Gene ID: (Human) 3996
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support