Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53268. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Plays a role in cell-cell adhesion and neuron guidance via its interactions with FLRT2 and FLRT3 that are expressed at the surface of adjacent cells. Plays a role in the development of glutamatergic synapses in the cortex. Important in determining the connectivity rates between the principal neurons in the cortex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adhesion G protein-coupled receptor L3; Calcium-independent alpha-latrotoxin receptor 3; CIRL-3; latrophilin homolog 3 (cow); Latrophilin-3; lectomedin 3; Lectomedin-3; Lph3
Gene Aliases: ADGRL3; CIRL3; CL3; KIAA0768; LEC3; LPHN3
UniProt ID: (Human) Q9HAR2
Entrez Gene ID: (Human) 23284
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support