Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (75%), Rat (75%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57505. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Component of the nexin-dynein regulatory complex (N-DRC) a key regulator of ciliary/flagellar motility which maintains the alignment and integrity of the distal axoneme and regulates microtubule sliding in motile axonemes. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dynein regulatory complex subunit 3; leucine rich repeat containing 48; Leucine-rich repeat-containing protein 48
Gene Aliases: CFAP134; DRC3; LRRC48
UniProt ID: (Human) Q9H069
Entrez Gene ID: (Human) 83450
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support