Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57469. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The NDUFA2 gene encodes one of the accessory subunits of complex I, the first and largest complex of the mitochondrial respiratory chain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CCDC76; CI-B8; complex I B8 subunit; Complex I-B8; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 2, 8kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 2; NADH-ubiquinone oxidoreductase B8 subunit; NADH-ubiquinone oxidoreductase subunit CI-B8
Gene Aliases: B8; CD14; CIB8; NDUFA2
UniProt ID: (Human) O43678
Entrez Gene ID: (Human) 4695
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support