Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Highest antigen sequence indentity to the following orthologs: Mouse (82%), Rat (82%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56079. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
NOL7 is encoded by this gene localizes to the nucleolus, where it maintains nucleolar structure and cell growth rates. The encoded protein also functions as a tumor suppressor and regulator of angiogenesis. The RB tumor suppressor gene recruits transcription factors to this gene and positively regulates its expression. Two transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Nucleolar protein 7; nucleolar protein 7, 27kDa; Nucleolar protein of 27 kDa; polyglutamine binding protein 3; retinoic acid repressible protein; U3 small nucleolar RNA-associated protein NOL7; U3 snoRNA-associated protein NOL7
Gene Aliases: C6orf90; dJ223E5.2; NOL7; NOP27; PQBP3; RARG-1
UniProt ID: (Human) Q9UMY1
Entrez Gene ID: (Human) 51406
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support