Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Highest antigen sequence indentity to the following orthologs: Mouse (92%), Rat (92%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82643. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene is related to the Wolf-Hirschhorn syndrome candidate-1 gene and encodes a protein with PWWP (proline-tryptophan-tryptophan-proline) domains. The function of the protein has not been determined. Two alternatively spliced variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp667H044; FLJ20353; Histone-lysine N-methyltransferase NSD3; MGC126766; MGC142029; nuclear receptor binding SET domain protein 3; Nuclear SET domain-containing protein 3; Protein whistle; WHSC1-like 1 isoform 9 with methyltransferase activity to lysine; WHSC1-like protein 1; Wolf-Hirschhorn syndrome candidate 1-like protein 1
Gene Aliases: DC28; NSD3; pp14328; WHISTLE; WHSC1L1
UniProt ID: (Human) Q9BZ95
Entrez Gene ID: (Human) 54904
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support