Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (65%), Rat (65%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55433. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein tyrosine phosphatase domain-containing protein 1; protein tyrosine phosphatase PTP9Q22
Gene Aliases: PTP9Q22; PTPDC1
UniProt ID: (Human) A2A3K4
Entrez Gene ID: (Human) 138639
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support