Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (79%), Rat (79%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84415. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Has transcriptional repressor activity. Overexpression inhibits the transcriptional activities of AP-1, p53/TP53 and CDKN1A. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hSmaug2; Protein Smaug homolog 2; SAM domain-containing protein 4B; Smaug 2; smaug homolog B; Sterile alpha motif domain-containing protein 4B
Gene Aliases: SAMD4B; SMAUG2; SMGB
UniProt ID: (Human) Q5PRF9
Entrez Gene ID: (Human) 55095
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support