Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (90%), Rat (90%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57380. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
SERINC1, also named as KIAA1253, TDE1L and TDE2, belongs to the TDE1 family. SERINC1 enhances the incorporation of serine into phosphatidylserine and sphingolipids.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Serine incorporator 1; Tumor differentially expressed protein 1-like; Tumor differentially expressed protein 2
Gene Aliases: KIAA1253; SERINC1; TDE1L; TDE2; TMS-2; TMS2; UNQ396/PRO732
UniProt ID: (Human) Q9NRX5
Entrez Gene ID: (Human) 57515
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support