Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (67%), Rat (67%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55897. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
UBXN10 (VCP/p97) binding protein required for ciliogenesis. Acts as a tethering factor that facilitates recruitment of VCP/p97 to the intraflagellar transport complex B (IFT-B) in cilia. UBX domain-containing proteins act as tethering factors for VCP/p97 and may specify substrate specificity of VCP/p97.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: UBX domain containing 3; UBX domain-containing protein 10; UBX domain-containing protein 3
Gene Aliases: UBXD3; UBXN10
UniProt ID: (Human) Q96LJ8
Entrez Gene ID: (Human) 127733
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support