Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63184. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
May participate in a protein complex showing an E3 ligase activity regulated by RAC1. Ubiquitination is directed towards itself and possibly other substrates, such as SMARCD2/BAF60b. Intrinsic E3 ligase activity has not been proven.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Putative E3 ubiquitin-protein ligase UNKL; RING finger protein unkempt-like; unkempt family zinc finger-like; unkempt homolog-like; Zinc finger CCCH domain-containing protein 5-like
Gene Aliases: C16orf28; UNKL; ZC3H5L; ZC3HDC5L
UniProt ID: (Human) Q9H9P5
Entrez Gene ID: (Human) 64718
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support