Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recombinant Human VEGF-C is a disulfide-linked homodimeric protein consisting of two 13.5 kDa polypeptide chains of 116 amino acid residues. Due to glycosylation, the protein migrates as a 30.0-33.0 kDa band by SDS-PAGE analysis under non-reducing conditions.
This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
The protein encoded by this gene is a member of the platelet-derived growth factor/vascular endothelial growth factor (PDGF/VEGF) family, is active in angiogenesis and endothelial cell growth, and can also affect the permeability of blood vessels. This secreted protein undergoes a complex proteolytic maturation, generating multiple processed forms which bind and activate VEGFR-3 receptors. Only the fully processed form can bind and activate VEGFR-2 receptors. This protein is structurally and functionally similar to vascular endothelial growth factor D.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Flt4 ligand; FLT4 ligand DHM; Flt4-L; Vascular endothelial growth factor C; Vascular endothelial growth factor-related protein; VEGF-C; VEGF3; VRP
Gene Aliases: Flt4-L; LMPH1D; VEGFC; VRP
UniProt ID: (Human) P49767
Entrez Gene ID: (Human) 7424
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support