Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Highest antigen sequence indentity to the following orthologs: Mouse (48%), Rat (48%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84416. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The function and protein-coding potential of this gene is unknown. The exon combination is based on AB058778.1 for which two possible open reading frames can be predicted (with start codons at nucleotide 26 or 2614). The position of the first ORF stop codon is consistent with a prediction of nonsense-mediated decay. Given the observation of the first ORF and its length, a predicted translation of the second ORF is inconsistent with the translation leaky scanning theory. Therefore, this gene is represented as a non-protein-coding transcript. [provided by RefSeq, Oct 2008].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: WD repeat-containing protein 97
Gene Aliases: KIAA1875; WDR97
UniProt ID: (Human) A6NE52
Entrez Gene ID: (Human) 340390
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support